ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4687.
 :  Avantor
 :  
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4687.
 :  Avantor
 :  
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501
  • For Laboratory,Research,or Manufacturing Use:
    0.0998-0.1002
  • Normality (by titrimetry):
    Passes Test
 :
 :40 g/mol
 :∼105 °C (1013 hPa)
 :1.327 g/cm³ (20 °C)
 :Ambient
 :8,II
 :Management of Change (MOC) category = N
 :215-185-5
 :1824
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548