Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sodium chloride concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32064.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32064.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (20°C):
    0.0995 - 0.1005 mol/l

Specification Test Results

About this item

 :
 :58.44 g/mol
 :Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477