Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Tetrachloromethane 100 µg/ml in methanol
Organic reference standard, Tetrachloromethane 100 µg/ml in methanol
Catalog # CMPLE798.D1.1ML.1
CAS Number:  
Organic reference standard, Tetrachloromethane 100 µg/ml in methanol
Catalog # CMPLE798.D1.1ML.1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1,5 mL
  • CAS No.:
    56-23-5
  • Description:
    Tetrachloromethane 100 µg/ml in methanol
  • Formula:
    CCl₄
  • Description:
    Tetrachloromethane

Specifications

About this item