Order Entry
Belgium
ContactUsLinkComponent
Osmium tetraoxide 4% in aqueous solution
Toxic
Osmium tetraoxide 4% in aqueous solution
Catalog # ROTH8088.1
Supplier:  Roth Carl
CAS Number:  
Toxic
Osmium tetraoxide 4% in aqueous solution
Catalog # ROTH8088.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

About this item

 :
 :254.23 g/mol
Storage Temperature:Refrigerator
EINECS:244-058-7
UN:3287
ADR:6.1,II
 :GHS06
CAS Number:20816-12-0
Formula:O₄Os
MDL Number:MFCD00011150