Order Entry
Belgium
ContactUsLinkComponent
 
 
Sulphuric acid solution 49-51% (by titrimetric analysis) for HPLC, Fluka™
 
 
Sulphuric acid solution 49-51% (by titrimetric analysis) for HPLC, Fluka™

 

  • Assay:
    49.0 - 51.0 %
  • Density (D 20/4):
    1.375 - 1.405
  • Absorbance 500 nm:
    Max. 0.02
  • Absorbance at 240 nm:
    Max. 0.02
  • Absorbance at 230 nm:
    Max. 0.03
  • Absorbance at 220 nm:
    Max. 0.04
  • Absorbance at 210 nm:
    Max. 0.05
  • Suitability test (HPLC):
    complying

 

 

Frequently Bought Together