Order Entry
Belgium
ContactUsLinkComponent
Sodium thiosulphate 0,1% in aqueous solution for microbiology
Sodium thiosulphate 0,1% in aqueous solution for microbiology
Catalog # AX011168
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium thiosulphate 0,1% in aqueous solution for microbiology
Catalog # AX011168
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50
  • Seq:
    0050
  • Pk Info:
    50×18 ml

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499