Order Entry
Belgium
ContactUsLinkComponent
Orthophosphoric acid ∼85%, TraceSELECT™ for trace analysis, Fluka™
Orthophosphoric acid ∼85%, TraceSELECT™ for trace analysis, Fluka™
Certificates
Orthophosphoric acid ∼85%, TraceSELECT™ for trace analysis, Fluka™

Specification Test Results

  • Assay:
    85.0 - 90.0 %
  • Density (20°C):
    1.691 - 1.706 g/ml
  • Silver (Ag):
    Max. 20 ppb
  • Aluminium (Al):
    Max. 20 ppb
  • Arsenic (As):
    Max. 0.005 ppm
  • Gold (Au):
    Max. 20 ppb
  • Barium (Ba):
    Max. 20 ppb
  • Beryllium (Be):
    Max. 20 ppb
  • Bismuth (Bi):
    Max. 20 ppb
  • Calcium (Ca):
    Max. 100 ppb
  • Cadmium (Cd):
    Max. 20 ppb
  • Cerium (Ce):
    Max. 20 ppb
  • Cobalt (Co):
    Max. 20 ppb
  • Chromium (Cr):
    Max. 50 ppb
  • Cesium (Cs):
    Max. 20 ppb
  • Copper (Cu):
    Max. 20 ppb
  • Iron (Fe):
    Max. 200 ppb
  • Gallium (Ga):
    Max. 20 ppb
  • Germanium (Ge):
    Max. 20 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 20 ppb
  • Iridium (Ir):
    Max. 20 ppb
  • Potassium (K):
    Max. 50 ppb
  • Lithium (Li):
    Max. 20 ppb
  • Magnesium (Mg):
    Max. 20 ppb
  • Manganese (Mn):
    Max. 20 ppb
  • Molybdenum (Mo):
    Max. 20 ppb
  • Sodium (Na):
    Max. 100 ppb
  • Nickel (Ni):
    Max. 50 ppb
  • Lead (Pb):
    Max. 20 ppb
  • Palladium (Pd):
    Max. 20 ppb
  • Platinum (Pt):
    Max. 20 ppb
  • Rubidium (Rb):
    Max. 20 ppb
  • Rhodium (Rh):
    Max. 20 ppb
  • Ruthenium (Ru):
    Max. 20 ppb
  • Antimony (Sb):
    Max. 500 ppb
  • Selenium (Se):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 20 ppb
  • Strontium (Sr):
    Max. 20 ppb
  • Titanium (Ti):
    Max. 20 ppb
  • Thallium (Tl):
    Max. 20 ppb
  • Uran (U):
    Max. 20 ppb
  • Vanadium (V):
    Max. 20 ppb
  • Zinc (Zn):
    Max. 20 ppb
  • Zirconium (Zr):
    Max. 20 ppb
  • Chloride (Cl):
    Max. 1 ppm
  • Nitrate (NO₃):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Ammonium (NH₄):
    Max. 1 ppm
  • KMnO₄ reducing matter (as O):
    Max. 1 ppm

Specification Test Results

Related Information

Frequently Bought Together