Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for schoolcap 802
Accessories for schoolcap 802
Accessories for schoolcap 802
Catalog # 135-1268
Supplier:
ERLAB CAPTAIRE
Expand image
Accessories for schoolcap 802
Catalog # 135-1268
Supplier:
ERLAB CAPTAIRE
Specifications
Description:
Waterproof box HCL for the storage of corrosive acids
Seq:
03
Pk:
1
Specifications
About this item
Product Details
Product Details