Orders LinkContactUsLinkComponent
Membrane filters, MembraClear, Whatman™
Membrane filters, MembraClear, Whatman™
Membrane filters, MembraClear, Whatman™
Certificates
Membrane filters, MembraClear, Whatman™
Membrane filters, MembraClear, Whatman™
2 Options Available Below
Product Details & Documents
These mixed cellulose ester membrane filters are designed for asbestos sampling using the membrane filter method for phase contrast microscopy. Asbestos sampling isolates these fibres from circulating air to determine concentrations.
  • Designed for use with the 'hot block method'
  • White with black grid for high contrast during asbestos testing using microscopy
  • Becomes transparent with acetone vapour treatment, distinguishing fibres from crystalline artifacts
Product Family Options
Product Information
  • Type
    Pore size
    Ø
    Availability
    Price
  • White with black grid
    0,80 µm
    25 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 176,00 €
    176,00 €
    1 * 100 ST
       176,00 €
  • White with black grid
    0,80 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 191,00 €
    191,00 €
    1 * 100 ST
       191,00 €