Order Entry
Belgium
ContactUsLinkComponent
Orthophosphoric acid ≥85%, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Orthophosphoric acid ≥85%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Orthophosphoric acid ≥85%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 85.0 %
  • Alkali phosphates:
    passes test
  • Color (APHA):
    max. 10
  • Chloride (Cl):
    max. 1 ppm
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.001 %
  • Nitrate (NO3):
    max. 5 ppm
  • Phosphorous or Hypophosphorous Acid:
    passes test
  • Reducing Substances:
    passes test
  • Sulfate (SO4):
    max. 0.003 %
  • Volatile Acids (as CH3COOH):
    max. 0.001 %
  • Aluminium (Al):
    max. 10
  • Arsenic (As):
    max. 0.5
  • Calcium (Ca):
    max. 20
  • Cadmium (Cd):
    max. 5
  • Copper (Cu):
    max. 5
  • Iron (Fe):
    max. 10
  • Potassium (K):
    max. 5
  • Magnesium (Mg):
    max. 10
  • Manganese (Mn):
    max. 0.5
  • Sodium (Na):
    max. 10
  • Nickel (Ni):
    max. 5
  • Lead (Pb):
    max. 5
  • Antimony (Sb):
    max. 4
  • Zinc (Zn):
    max. 10

Specification Test Results

Related Information

Frequently Bought Together