Orders LinkContactUsLinkComponent
Sample dippers
Sample dippers
Supplier:   Brand
Sample dippers
Supplier:  Brand
Certificates
Sample dippers
4 Options Available Below
Product Details & Documents
Sampling dipper with reinforced rim and spout.
  • PTFE handle with steel core
Optional extension rod for increased handle length.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Capacity
    Availability
    Price
  • BRND90438
    Sampling dipper with reinforced rim and spout
    Sampling dipper with reinforced rim and spout
    PTFE/steel
    65 mm
    55 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 168,00 €
    168,00 €
    1 * 1 ST
       168,00 €
  • BRND90448
    Sampling dipper with reinforced rim and spout
    Sampling dipper with reinforced rim and spout
    PTFE/steel
    95 mm
    70 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 205,00 €
    205,00 €
    1 * 1 ST
       205,00 €
  • BRND90454
    Sampling dipper with reinforced rim and spout
    Sampling dipper with reinforced rim and spout
    PTFE/steel
    125 mm
    80 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 237,00 €
    237,00 €
    1 * 1 ST
       237,00 €
  • BRND90462
    Sampling dipper with reinforced rim and spout
    Sampling dipper with reinforced rim and spout
    PTFE/steel
    155 mm
    105 mm
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 285,00 €
    285,00 €
    1 * 1 ST
       285,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extension rod for sampling device, 600 mm
    Extension rod for sampling device, 600 mm
    BRND90470
    Log in for availability
    1 * 1 ST - 121,00 €