Orthophosphoric acid 5%, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
Frequently Bought Together