Order Entry
Belgium
ContactUsLinkComponent
Orthophosphoric acid 10%, TECHNICAL
Corrosive
Orthophosphoric acid 10%, TECHNICAL
Catalog # 31142.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Orthophosphoric acid 10%, TECHNICAL
Catalog # 31142.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 10 %
  • Appearance:
    Clear colourless liquid

Specification Test Results

About this item

 :
 :98 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340