Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 35% in aqueous solution, GPR RECTAPUR®
Corrosive
Sodium hydroxide 35% in aqueous solution, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sodium hydroxide 35% in aqueous solution, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    34 - 36 %
  • Density (20/4):
    1.370 - 1.390
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together