Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Corrosive
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 32 %
  • Total N (Nitrogen):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together