Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide ∼32% in aqueous solution, EMPLURA®, Supelco®
Corrosive
Sodium hydroxide ∼32% in aqueous solution, EMPLURA®, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Sodium hydroxide ∼32% in aqueous solution, EMPLURA®, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    9 °C
  • Boiling Point:
    119 °C (1013 hPa)
  • Vapor Pressure:
    8 hPa (20 °C)
  • Density:
    1.35 g/cm3 (20 °C)
  • pH:
    14 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Frequently Bought Together