Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide ≥32% in aqueous solution, Purum, Fluka™
Corrosive
Sodium hydroxide ≥32% in aqueous solution, Purum, Fluka™
Catalog # 05211-2.5L
CAS Number:  
Corrosive
Sodium hydroxide ≥32% in aqueous solution, Purum, Fluka™
Catalog # 05211-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    Min. 32 %
  • Iron (Fe):
    Max. 0.002 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:120 °C
Melting Pt:9…10 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548