Order Entry
Belgium
ContactUsLinkComponent
Hydrogen peroxide 35%, Supelco®
Corrosive
Irritant
Hydrogen peroxide 35%, Supelco®
Catalog # 1.08556.9025
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Hydrogen peroxide 35%, Supelco®
Catalog # 1.08556.9025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -24 °C
  • Boiling Point:
    110 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Density:
    1.13 g/cm3 (20 °C)
  • pH:
    2 - 4 (H2O, 20 °C)

Specification Test Results

About this item

 :
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2014
ADR:5.1 (8),II
 :M-Clarity™ quality level = MQ200
 :34.01 g/mol
EINECS:231-765-0
 :GHS05
 :GHS07
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333