Order Entry
Belgium
ContactUsLinkComponent
Hydrogen peroxide 3% stabilised, GPR RECTAPUR®
Hydrogen peroxide 3% stabilised, GPR RECTAPUR®
Catalog # 23614.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrogen peroxide 3% stabilised, GPR RECTAPUR®
Catalog # 23614.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    2.5 - 3.5 %
  • Evaporation residue:
    Max. 0.05 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • Fe (Iron):
    Max. 5 ppm

Specification Test Results

About this item

 :
Boiling Pt:100 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
 :Stabilised with phosphate
 :34.01 g/mol
Storage Temperature:Refrigerator
EINECS:231-765-0
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333