Nitric acid 65%, EMSURE® ISO for analysis (max. 0.005 ppm Hg), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8 (5.1),II
:M-Clarity™ quality level = MQ300
EINECS:231-714-2
:GHS03
:GHS06
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-32 °C
- Boiling Point:121 °C (1013 hPa)
- Vapor Pressure:9.4 hPa (20 °C)
- Density:1.39 g/cm³ (20 °C)
- pH:(H₂O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00452.1000_TDS_EN ProductPage_100452
Product specification
msds_100452
SDS
Frequently Bought Together