Order Entry
Belgium
ContactUsLinkComponent
Hydrofluoric acid 48%, Ultrapure for trace analysis, Supelco®
Toxic
Corrosive
Hydrofluoric acid 48%, Ultrapure for trace analysis, Supelco®
Catalog # 1.01513.1000
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 48%, Ultrapure for trace analysis, Supelco®
Catalog # 1.01513.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :20.01 g/mol
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ100
EINECS:231-634-8
UN:1790
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346