Order Entry
Belgium
ContactUsLinkComponent
Hydrofluoric acid 48 - 51% ≥98% ACS
Toxic
Corrosive
Hydrofluoric acid 48 - 51% ≥98% ACS
Catalog # 33258.09
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 48 - 51% ≥98% ACS
Catalog # 33258.09
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :20.01 g/mol
Merck Index:14,4790
Boiling Pt:112 °C
Density:1.16
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS06
 :GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346