Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Hydrofluoric acid 40%, GPR RECTAPUR®
  20306.296
 :  
undefined
 
 
Hydrofluoric acid 40%, GPR RECTAPUR®
  20306.296
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    40 - 42 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm

 

 

 :
 :20.01 g/mol
 :112 °C (1013 hPa)
 :–44 °C
 :Ambient
 :231-634-8
 :1790
 :8 (6.1),II
 :GHS06
 :GHS05
 :7664-39-3
 :HF
 :MFCD00011346