Order Entry
Belgium
ContactUsLinkComponent
Hydrofluoric acid 40%, ARISTAR® for trace analysis
Hydrofluoric acid 40%, ARISTAR® for trace analysis
Catalog # 450034N
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrofluoric acid 40%, ARISTAR® for trace analysis
Catalog # 450034N
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay (acidimetric):
    Min. 40.0 %
  • Colour:
    Max. 10 APHA
  • Cl (Chloride):
    Max. 0.5 ppm
  • SiF6 (Hexafluorosilicate):
    Max. 20 ppm
  • PO4 (Phosphate):
    Max. 0.1 ppm
  • SO4 (Sulphate):
    Max. 0.5 ppm
  • SO3 (Sulphite):
    Max. 2 ppm
  • Ag (Silver):
    Max. 0.001 ppm
  • Al (Aluminium):
    Max. 0.005 ppm
  • Au (Gold):
    Max. 0.002 ppm
  • As (Arsenic):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.010 ppm
  • Be (Beryllium):
    Max. 0.001 ppm
  • Bi (Bismuth):
    Max. 0.001 ppm
  • Ca (Calcium):
    Max. 0.010 ppm
  • Cd (Cadmium):
    Max. 0.002 ppm
  • Co (Cobalt):
    Max. 0.002 ppm
  • Cr (Chromium):
    Max. 0.001 ppm
  • Cu (Copper):
    Max. 0.001 ppm
  • Fe (Iron):
    Max. 0.010 ppm
  • Ga (Gallium):
    Max. 0.005 ppm
  • Ge (Germanium):
    Max. 0.002 ppm
  • Hg (Mercury):
    Max. 0.020 ppm
  • In (Indium):
    Max. 0.002 ppm
  • K (Potassium):
    Max. 0.010 ppm
  • Li (Lithium):
    Max. 0.001 ppm
  • Mg (Magnesium):
    Max. 0.005 ppm
  • Mn (Manganese):
    Max. 0.002 ppm
  • Mo (Molybdenum):
    Max. 0.001 ppm
  • Na (Sodium):
    Max. 0.010 ppm
  • Ni (Nickel):
    Max. 0.001 ppm
  • Pb (Lead):
    Max. 0.002 ppm
  • Sn (Tin):
    Max. 0.001 ppm
  • Sr (Strontium):
    Max. 0.001 ppm
  • Ti (Titanium):
    Max. 0.010 ppm
  • Tl (Thallium):
    Max. 0.001 ppm
  • V (Vanadium):
    Max. 0.001 ppm
  • Zn (Zinc):
    Max. 0.005 ppm
  • Zr (Zirconium):
    Max. 0.001 ppm
  • Ignition residue (SO4):
    Max. 2 ppm

Specification Test Results

About this item

 :
 :20.01 g/mol
Boiling Pt:112 °C (1013 hPa)
Melting Pt:–44 °C
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8 (6.1),II
 :GHS06
 :GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346