Orders LinkContactUsLinkComponent
OmniTrays, Nunclon™Δ, with enhanced rectangular area
OmniTray, enhanced rectangular area, Clear, 84 cm², 90 ml
  734-2402
   Low Manufacturing Impact
OmniTrays, Nunclon™Δ, with enhanced rectangular area
  734-2402
   Low Manufacturing Impact
OmniTrays, Nunclon™Δ, with enhanced rectangular area
OmniTrays, Nunclon™Δ, with enhanced rectangular area
OmniTray, enhanced rectangular area, Clear, 84 cm², 90 ml
665,00 €
1 * 90 ST
PS, sterile. Optimised for computerised imaging and spotting.
  • Colour:
    Clear
  • Description:
    OmniTray, enhanced rectangular area
  • Culture area:
    84 cm²
  • Total volume:
    90 ml
  • Cat. No.:
    734-2402
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact