Nalgene® Media Bottles, Square, Amber, with Screw Cap
PETG, amber, translucent, with HDPE screw cap.
- Non-sterile
- Non-pyrogenic
- Non-cytotoxic
Ideal for storage and shipping of light sensitive liquid media, buffers, and sera. Heavy-walled bottle provides an excellent gas barrier; features molded-in graduations, with a drip-proof neck finish and molded-in neck ring.
Heavy-walled, shatter resistant and durable, space-saving square design, molded in graduations, nonsterile, non-pyrogenic, non-cytotoxic, leakproof, with drip-proof neck finish and molded-in neck ring, PETG provides excellent gas barrier, with transparency and reduced permeability to CO₂ and O₂, amber bottle and closure provide protection for light sensitive reagents.
: Meets current standards for USP class VI, USP 661, and european pharmacopeia for abnormal toxicity. PETG media bottles and HDPE closure materials meet current biological testing standards including USP class VI.
- Catalog No:
- NALG322021-0125
- NALG322021-0030
- NALG322021-0060
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together