Pipette tips, microvolume, Corning®
Supplier: Corning
About this item
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
4 Options Available Below
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- 613-4242
- 613-4245
- 734-1987
- 613-5516
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
- Tip styleVolumeSterileTip colourFor use withPackedAvailabilityPrice
- Round tip0,1 - 10 µl-TransparentGilson®, other Ultra-Micropipettors10 racks, 96 eachLog in for availability1 * 960 ST94,70 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.613-4242Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Round tip0,1 - 10 µl-TransparentGilson, other Ultra-MicropipettorsBulk; 10000 tips/bagLog in for availability1 * 10.000 ST496,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.613-4245Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Round tip0,1 - 10 µl+TransparentGilson, other Ultra-Micropipettors10 racks, 96 eachLog in for availability1 * 960 ST118,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1987Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - Round tip0,5 - 10 µl-TransparentEppendorf, other Ultra-MicropipettorsBulk; 10000 tips/bagLog in for availability1 * 10.000 ST444,00 €Specifications:
More
Specifications
Cat. No.613-5516Environmentally PreferableEnvironmental certification