Order Entry
Belgium
ContactUsLinkComponent
 
Perchloric acid ≥70-72%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.00519.1001
Supplier:  Merck
CAS Number:  
undefined
Perchloric acid ≥70-72%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.00519.1001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -18 °C
  • Boiling Point:
    198.7 °C (1013 hPa)
  • Density:
    1.68 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1 (8),I
 :M-Clarity™ quality level = MQ300
 :GHS03
 :GHS05
 :GHS08
 :GHS07
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325