Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 6 mol/l (6 N), AVS TITRINORM Reag. Ph. Eur. 3001500 volumetric solution
Hydrochloric acid 6 mol/l (6 N), AVS TITRINORM Reag. Ph. Eur. 3001500 volumetric solution
Catalog # 2611.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid 6 mol/l (6 N), AVS TITRINORM Reag. Ph. Eur. 3001500 volumetric solution
Catalog # 2611.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    5.976 - 6.024 mol/l

Specification Test Results

About this item

 :
Density:1.06 g/cm³ (20 °C)
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324