Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hydrochloric acid 37%

Hydrochloric acid 37%

Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Hydrochloric acid 37%
Supplier:  Roth Carl
Marketsource
Certificates
Hydrochloric acid 37%

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
    Marketsource
    Marketsource