

Hydrochloric acid 30%, Ultrapure for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (acidimetric):≥ 30.0 %
- Ag (Silver):≤ 100 ppt
- Al (Aluminium):≤ 100 ppt
- As (Arsenic):≤ 100 ppt
- Au (Gold):≤ 100 ppt
- B (Boron):≤ 100 ppt
- Ba (Barium):≤ 50 ppt
- Be (Beryllium):≤ 50.0 ppt
- Bi (Bismuth):≤ 50.0 ppt
- Ca (Calcium):≤ 500 ppt
- Cd (Cadmium):≤ 50.0 ppt
- Ce (Cerium):≤ 20.00 ppt
- Co (Cobalt):≤ 100.0 ppt
- Cr (Chromium):≤ 50 ppt
- Cu (Copper):≤ 100 ppt
- Dy (Dysprosium):≤ 20.00 ppt
- Er (Erbium):≤ 20.00 ppt
- Eu (Europium):≤ 20.00 ppt
- Fe (Iron):≤ 500 ppt
- Ga (Gallium):≤ 50.0 ppt
- Gd (Gadolinium):≤ 20 ppt
- Hf(Hafnium):≤ 10.00 ppt
- Hg (Mercury):≤ 50 ppt
- Ho (Holmium):≤ 20.00 ppt
- In (Indium):≤ 50.0 ppt
- K (Potassium):≤ 500 ppt
- La (Lanthanum):≤ 20.0 ppt
- Li (Lithium):≤ 50.0 ppt
- Lu (Lutetium):≤ 20.00 ppt
- Mg (Magnesium):≤ 100 ppt
- Mn (Manganese):≤ 50 ppt
- Mo (Molybdenum):≤ 50.0 ppt
- Na (Sodium):≤ 200 ppt
- Nb (Niobium):≤ 20.0 ppt
- Nd (Neodymium):≤ 20.0 ppt
- Ni (Nickel):≤ 200 ppt
- Pb (Lead):≤ 50.0 ppt
- Pd (Palladium):≤ 100 ppt
- Pr (Praseodym):≤ 20.00 ppt
- Pt (Platinum):≤ 50 ppt
- Rb (Rubidium):≤ 20.0 ppt
- Re (Rhenium):≤ 10.00 ppt
- Ru (Ruthenium):≤ 20.0 ppt
- Sb (Antimony):≤ 100 ppt
- Sc (Scandium):≤ 20 ppt
- Se (Selenium):≤ 200 ppt
- Si (Silicon):≤ 1000 ppt
- Sm (Samarium):≤ 20.00 ppt
- Sn (Tin):≤ 200 ppt
- Sr (Strontium):≤ 50.0 ppt
- Ta (Tantalum):≤ 100.00 ppt
- Tb (Terbium):≤ 20.00 ppt
- Te (Tellur):≤ 100 ppt
- Th (Thorium):≤ 1.00 ppt
- Ti (Titanium):≤ 50 ppt
- Tl (Thallium):≤ 50.0 ppt
- Tm (Thulium):≤ 20.00 ppt
- U (Uranium):≤ 1.00 ppt
- V (Vanadium):≤ 50 ppt
- W (Wolfram):≤ 100 ppt
- Y (Yttrium):≤ 10.0 ppt
- Yb (Ytterbium):≤ 20.0 ppt
- Zn (Zinc):≤ 200 ppt
- Zr (Zirconium):≤ 20.0 ppt
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure Merck Ultrapur and Suprapur® High Purity Acids and Bases
Product Literature
msds_101514
SDS
ProductPage_101514
Product specification
Frequently Bought Together