Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 25%, EMSURE® for analysis, Supelco®
Corrosive
Irritant
Hydrochloric acid 25%, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Irritant
Hydrochloric acid 25%, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (Alkalimetric):
    ≥25.0%
  • Color:
    ≤10 Hazen
  • Bromide (Br):
    ≤50 ppm
  • Free Chlorine (Cl):
    ≤0.5 ppm
  • Sulfate (SO₄):
    ≤1 ppm
  • Sulfite (SO₃):
    ≤1 ppm
  • Al (Aluminium):
    ≤0.050 ppm
  • As (Arsenic):
    ≤0.01 ppm
  • Ba (Barium):
    ≤0.020 ppm
  • Be (Beryllium):
    ≤0.020 ppm
  • Bi (Bismuth):
    ≤0.100 ppm
  • Ca (Calcium):
    ≤0.500 ppm
  • Cd (Cadmium):
    ≤0.010 ppm
  • Co (Cobalt):
    ≤0.010 ppm
  • Cr (Chromium):
    ≤0.020 ppm
  • Cu (Copper):
    ≤0.020 ppm
  • Fe (Iron):
    ≤0.200 ppm
  • Ge (Germanium):
    ≤0.050 ppm
  • K (Potassium):
    ≤0.100 ppm
  • Li (Lithium):
    ≤0.010 ppm
  • Mg (Magnesium):
    ≤0.100 ppm
  • Mn (Manganese):
    ≤0.010 ppm
  • Mo (Molybdenum):
    ≤0.020 ppm
  • NH₄ (Ammonium):
    ≤1 ppm
  • Na (Sodium):
    ≤0.500 ppm
  • Ni (Nickel):
    ≤0.020 ppm
  • Pb (Lead):
    ≤0.020 ppm
  • Sr (Strontium):
    ≤0.010 ppm
  • Ti (Titanium):
    ≤0.100 ppm
  • Tl (Thallium):
    ≤0.050 ppm
  • V (Vanadium):
    ≤0.010 ppm
  • Zn (Zinc):
    ≤0.050 ppm
  • Zr (Zirconium):
    ≤0.100 ppm
  • Residue on Ignition (as SO₄):
    ≤5 ppm

Specification Test Results

Frequently Bought Together