Hydrochloric acid 20%
Glass bottle for solids, 500 g
Catalog # WAKO632-03305
Marketsource
Supplier: Wako
Jump to:
Product Details & Documents
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46 g/mol
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



