Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Proteomics Grade
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Proteomics Grade
Catalog # M112-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Proteomics Grade
Catalog # M112-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • DNase:
    NONE
  • pH @25 °C:
    5.0 – 8.0
  • Protease:
    NONE
  • RNase:
    NONE

Specification Test Results

About this item

 :
 :288.38 g/mol
Density:1…1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,
 :GHS07
 :GHS05
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175