Order Entry
Belgium
ContactUsLinkComponent
 
 
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution, Ultra Pure Grade
  E719-100ML
 :  
undefined
 
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution, Ultra Pure Grade
  E719-100ML
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Color (APHA):
    <5

 

 

 :
 :288.38 g/mol
 :1…1.1 g/cm³ (20 °C)
 :205-788-1
 :GHS05
 :Ambient
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175