


Trichloroacetic acid (TCA) 200 g/l in aqueous solution
Catalog # PRLSMC313064.1000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:163.39 g/mol
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:≤1.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-927-2
UN:2564
ADR:8,II
:GHS05
:GHS07
:GHS09
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177