Order Entry
Belgium
ContactUsLinkComponent
Formic acid, EMSURE® ACS for analysis, Supelco®
Toxic
Corrosive
Formic acid, EMSURE® ACS for analysis, Supelco®
Catalog # 1.00253.1000
Supplier:  Merck
CAS Number:  
undefined
Toxic
Corrosive
Formic acid, EMSURE® ACS for analysis, Supelco®
Catalog # 1.00253.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -9 °C
  • Boiling Point:
    107 °C
  • Flash Point:
    71 °C
  • Explosion Limit:
    10 - 45.5 %(V)
  • Density:
    1.2 g/cm3 (20 °C)
  • pH:
    (H2O) strongly acid
  • Ignition Point:
    485 °C

Specification Test Results

About this item

 :
 :46.03 g/mol
Merck Index:13,04265
ADR:8 (3),II
 :M-Clarity™ quality level = MQ300
EINECS:200-579-1
UN:1779
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297