


Formaldehyde 37% in aqueous solution stabilised, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
ADR:8,III
:Stabilised with methanol
EINECS:200-001-8
UN:2209
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:37.0 - 38.0 %
- Acidity:passes test
- Appearance of solution:passes test
- Identification:passes test
- Methanol (CH3OH):9.0 - 15.0 %
- Sulfated Ash:max. 0.1 %
Specification Test Results
Frequently Bought Together