


Formaldehyde 37% in aqueous solution stabilised, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
ADR:8,III
:Stabilised with methanol
EINECS:200-001-8
UN:2209
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:36.5 - 38.0 %
- Color (APHA):max. 10
- Preservative (CH3OH):10 - 15 %
- Residue after Ignition:max. 0.01 %
- Sulfate (SO4):max. 0.002 %
- Titrable Acid (meq/g):max. 0.01
- Chloride (Cl):max. 5
- Iron (Fe):max. 5
- Heavy Metals (as Pb):max. 5
Specification Test Results
Frequently Bought Together