Order Entry
Belgium
ContactUsLinkComponent
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
Toxic
Health
Corrosive
Flammable
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
Certificates
undefined
Toxic
Health
Corrosive
Flammable
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    10 to 15 % methanol
  • Residue after ignition:
    ≤0.005 %
  • Titration:
    36.5 to 38.0 %
  • Chloride (Cl):
    ≤5 ppm
  • Sulfate (SO4):
    ≤0.002 %
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (ICP-OES):
    ≤5 ppm
  • Titrable acid:
    ≤0.006 meq/g

Specification Test Results

Related Information

Frequently Bought Together