Order Entry
Belgium
ContactUsLinkComponent
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Health
Toxic
Corrosive
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Toxic
Corrosive
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    35 - 38 %
  • Density (20/4):
    1.083 - 1.093
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Methanol:
    8 - 12 %

Specification Test Results

Frequently Bought Together