Formaldehyde 35% in aqueous solution
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
4 Options Available Below
Product Details & Documents
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



