Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
   Sustainable Options Available
Dropping bottles, LDPE, Nalgene®
   Sustainable Options Available
Certificates
Dropping bottles, LDPE, Nalgene®
6 Options Available Below
Product Details & Documents
Dropper bottles made up of natural/white low density polyethylene.
  • Round shape
  • Drop size: 40/50 μl
  • Leakproof
Certifications: Compliance with ISO 13485:2003; 21CFR177.1520 and USP Class VI.
Ordering information: Controlled-drop dispensing tip and coloured closures are available seperately.
Specifications for Product Family
Specifications
  • Catalog No:
  • NALG312751-9125
  • 215-2350
  • NALG312751-9050
  • NALG312750-9125
  • NALG312750-9025
  • NALG312750-9050
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
White
Product Information
  • Capacity
    Material
    Height
    Ext.Ø
    Thread
    For
    Availability
    Price
  • 4 ml
    LDPE
    54 mm
    20 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 220,00 €
    1 220,00 €
    1 * 2.000 ST
       1 220,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG312751-9125
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 8 ml
    LDPE
    65 mm
    20 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 240,00 €
    1 240,00 €
    1 * 2.000 ST
       1 240,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-2350
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 15 ml
    LDPE
    75 mm
    25 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 440,00 €
    1 440,00 €
    1 * 2.000 ST
       1 440,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG312751-9050
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Natural
Product Information
  • Capacity
    Material
    Height
    Ext.Ø
    Thread
    For
    Availability
    Price
  • 4 ml
    LDPE
    54 mm
    20 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 200,00 €
    1 200,00 €
    1 * 2.000 ST
       1 200,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG312750-9125
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 8 ml
    LDPE
    65 mm
    20 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 240,00 €
    1 240,00 €
    1 * 2.000 ST
       1 240,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG312750-9025
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 15 ml
    LDPE
    75 mm
    25 mm
    15-415 mm
    Nalgene controlled dropper
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 1 420,00 €
    1 420,00 €
    1 * 2.000 ST
       1 420,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG312750-9050
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Closures, PP, yellow
    Closures, PP, yellow
    NALG312760-0020
    Log in for availability
    1 * 2.000 ST - 668,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Dispensing tips, LDPE, clear, 50 µl
    Dispensing tips, LDPE, clear, 50 µl
    NALG312758-0001
    Log in for availability
    1 * 2.000 ST - 476,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Dispensing tips, LDPE, clear, 40 µl
    Dispensing tips, LDPE, clear, 40 µl
    NALG312759-0001
    Log in for availability
    1 * 2.000 ST - 568,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Closures, PP, natural
    Closures, PP, natural
    NALG312760-0000
    Log in for availability
    1 * 2.000 ST - 668,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Closures, PP, white
    Closures, PP, white
    NALG312760-0010
    Log in for availability
    1 * 2.000 ST - 662,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Closures, PP, green
    Closures, PP, green
    NALG312760-0040
    Log in for availability
    1 * 2.000 ST - 668,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Closures, PP, blue
    Closures, PP, blue
    NALG312760-0060
    Log in for availability
    1 * 2.000 ST - 668,00 €