Centrifuge tubes, conical bottom, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, translucent.
- Excellent chemical resistance
- Maximum RCF: 6000 ×g
- Autoclavable
Conical-bottom centrifuge tubes, which feature molded-in graduations.
Specifications
- Catalog No:
- 525-0465
- 525-0464
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement