ContactUsLinkComponent
Methanol-[D₁] 99.0 atom % D for NMR spectroscopy
Toxic
Health
Flammable
Methanol-[D₁] 99.0 atom % D for NMR spectroscopy
Certificates
undefined
Toxic
Health
Flammable
Methanol-[D₁] 99.0 atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (Coulometric)
  • D-Enrichment:
    ≥99.0 %