

Xylene (mixture of isomers), anhydrous (max. 0.005% H₂O) ≥99.5% (xylenes + ethylbenzene basis), AnalaR NORMAPUR® analytical reagent
Catalog # 28976.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:Clear colourless liquid
- Colour value:Max. 10 APHA
- Assay (Xylene isomers + EB):Min. 99.5 %
- Residue on evaporation:Max. 0.0010 %
- Water (K.F.):Max. 0.0050 %
- Benzene:Max. 0.100 %
- Toluene:Max. 1 %
- Thiophene:Max. 0.00010 %
- Acidity or alkalinity:Max. 0.00020 meq/g
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.01 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.05 ppm
- Total S (Sulphur):Max. 5 ppm
Specification Test Results
About this item
:
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
REACH:01-2119488216-32
:GHS02
:GHS07
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264