Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Xylene (mixture of isomers), BAKER ANALYZED®, Laboratory Reagent for spectrophotometry, J.T.Baker®
  9516.1000
 :  Avantor
 :  
undefined
 
 
Xylene (mixture of isomers), BAKER ANALYZED®, Laboratory Reagent for spectrophotometry, J.T.Baker®
  9516.1000
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Boiling Range (initial to dry point):
    137 - 144 °C
  • Color (APHA):
    max. 10
  • Free Acid (µeq/g):
    max. 0.12
  • Residue after Evaporation:
    max. 0.001 %
  • Sulfur Compounds (as S):
    max. 0.003 %
  • Substances Darkened by H2SO4:
    passes test
  • Water (H2O):
    max. 0.02 %
  • at 295 nm:
    max. 1.00
  • at 300 nm:
    max. 0.30
  • at 330 nm:
    max. 0.05
  • at 380-400 nm:
    max. 0.01

 

 

 :
 :106.17 g/mol
 :13,10136
 :140 °C (1013 hPa)
 :–34 °C
 :0.86…0.88 g/cm³ (20 °C)
 :25 °C
 :Ambient
 :215-535-7
 :1307
 :3,III
 :GHS02
 :GHS07
 :1330-20-7
 :C₆H₄(CH₃)₂
 :MFCD00077264