Orders LinkContactUsLinkComponent
Closed dome tanks
   Sustainable Options Available
Closed dome tanks
   Sustainable Options Available
Certificates
Closed dome tanks
4 Options Available Below
Product Details & Documents
These dome tanks are designed for use as closed containment systems. Tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sealing.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • NALG2650-0020
  • NALG2650-0030
  • NALG2650-0055
  • NALG2650-0100
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ø ext.×H
    Availability
    Price
  • NALG2650-0020
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    75 l
    419×813 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 200,00 €
    1 200,00 €
    1 * 1 ST
       1 200,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG2650-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG2650-0030
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    115 l
    470×981 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 690,00 €
    1 690,00 €
    1 * 1 ST
       1 690,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG2650-0030
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG2650-0055
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    210 l
    559×1099 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG2650-0055
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG2650-0100
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    Closed dome tank, with silicone gasketed PP closure, neck opening: 150 mm
    380 l
    724×1321 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 520,00 €
    3 520,00 €
    1 * 1 ST
       3 520,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG2650-0100
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Cap Size
    For bottles
    Availability
    Price
  • with silicone gasket, true union clamp
    with silicone gasket, true union clamp
    NALG2651-0200
    152,3 mm
    75/115/ 210/380 l
    Log in for availability
    1 * 1 ST - 1 250,00 €