Order Entry
Belgium
ContactUsLinkComponent
 
Health
Irritant
Formaldehyde 10% in aqueous solution stabilised
Catalog # 5167.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Formaldehyde 10% in aqueous solution stabilised
Catalog # 5167.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (Formaldehyde):
    8 - 12 %
  • pH (20°C):
    8.2 - 8.4

Specification Test Results

About this item

 :
 :30.03 g/mol
ADR:9,
 :Stabilised with methanol 0,5-1,5 %
EINECS:200-001-8
UN:3334
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274