Orders LinkContactUsLinkComponent
Nalgene®, Biotainer® Bottles
Nalgene®, Biotainer® Bottles
   Options durables disponibles
Nalgene®, Biotainer® Bottles
   Options durables disponibles
Certificats
Nalgene®, Biotainer® Bottles
9 Options disponibles ci-dessous
Product Details & Documents
PETG, crystal clear.
  • No risk of valuable contents loss with the leakproof assurance
  • Puncture and impact resistant bottles secure your highly valuable contents
  • Supported by extensive validation documentation to reduce time and cost at implementation
  • Customised fluid transfer options available to fit your specific process application
  • Easy-to-read graduations, lot number and expiration date for convenient, no-mistake traceability
Biotainer™ bottles are designed specifically for the storage and transportation of active pharmaceutical ingredients. Extensive regulatory documentation assures that the products meet your demanding biotechnology requirements and reduces the time and cost to implement them in your process.
Eliminate the cost of cleaning, sterilising and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁶.
Durable, crystal clear PETG for sample integrity, closures with polyethylene foam core liners for leakproof assurance, custom options available for fluid transfer applications, supported by extensive validation documentation, suitable for storage in conditions from −40 to +70 °C.
Certifications: USP class VI, non-cytotoxic, USP <661>, non-pyrogenic, FDA 21CFR 177.1315.
Spécifications de la famille de produits
Spécifications
  • Catalog No:
  • NALG3025-42
  • NALG3005-42
  • NALG3005-70
  • NALG3110-35
  • NALG3110-42
  • NALG3230-42
  • NALG3230-20
  • NALG3415-16
  • NALG3415-42
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Spécifications
Product Family Options
Informations sur le produit
  • Capacity
    Type
    Thread
    Material
    Colour
    Neck I-Ø
    W×D×H
    Cap Size
    For
    Packed
    Availability
    Price
  • 125 ml
    with closure, sterile
    38 mm
    PETG
    Clear
    28 mm
    52×52×105 mm
    38 mm
    Prolonged storage
    5 per pack/100 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 100 ST
    1 * 100 ST - 844,00 €
    844,00 €
    1 * 100 ST
       844,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3025-42
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 500 ml
    with closure, sterile
    38 mm
    PETG
    Clear
    28 mm
    77×77×176 mm
    38 mm
    Prolonged storage
    5 per pack/70 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 70 ST
    1 * 70 ST - 725,00 €
    725,00 €
    1 * 70 ST
       725,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3005-42
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 500 ml
    with closure, sterile
    38 mm
    PETG
    Clear
    28 mm
    77×77×176 mm
    38 mm
    Prolonged storage
    35 per pack/70 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 70 ST
    1 * 70 ST - 686,00 €
    686,00 €
    1 * 70 ST
       686,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3005-70
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    with closure, sterile
    48 mm
    PETG
    Clear
    37 mm
    98×98×201 mm
    Prolonged storage
    35 per pack/35 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 35 ST
    1 * 35 ST - 638,00 €
    638,00 €
    1 * 35 ST
       638,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3110-35
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    with closure, sterile
    48 mm
    PETG
    Clear
    37 mm
    98×98×201 mm
    Prolonged storage
    5 per pack/35 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 35 ST
    1 * 35 ST - 678,00 €
    678,00 €
    1 * 35 ST
       678,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3110-42
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    with closure, sterile
    48 mm
    PETG
    Clear
    36 mm
    116×116×265 mm
    48 mm
    Prolonged storage
    5 per pack/20 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 20 ST
    1 * 20 ST - 475,00 €
    475,00 €
    1 * 20 ST
       475,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3230-42
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    with closure, sterile
    48 mm
    PETG
    Clear
    36 mm
    116×116×265 mm
    48 mm
    Prolonged storage
    20 per pack/20 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 20 ST
    1 * 20 ST - 458,00 €
    458,00 €
    1 * 20 ST
       458,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3230-20
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5000 ml
    with closure, sterile, with handle
    48 mm
    PETG
    Clear
    37 mm
    166×166×290 mm
    48 mm
    Prolonged storage
    1 per pack/6 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 6 ST
    1 * 6 ST - 559,00 €
    559,00 €
    1 * 6 ST
       559,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3415-16
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5000 ml
    with closure, sterile
    48 mm
    PETG
    Clear
    165 mm
    98×98×290 mm
    48 mm
    Prolonged storage
    6 per pack/6 per case
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 6 ST
    1 * 6 ST - 550,00 €
    550,00 €
    1 * 6 ST
       550,00 €
    Spécifications :
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG3415-42
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Thread
    Cap Size
    Availability
    Price
  • Biotainer® replacement closure, PP, sterile, with silicone liner
    Biotainer® replacement closure, PP, sterile, with silicone liner
    NALG362515-0480
    48 mm
    48 mm
    Connexion Pour la disponibilité
    1 * 300 ST - 3 750,00 €
Informations sur le produit
  • Catalog #
    Cap Size
    Type
    Availability
    Price
  • 3-ported closure, two ports 6,36 mm I.Ø., one port 4,76 mm
    3-ported closure, two ports 6,36 mm I.Ø., one port 4,76 mm
    NALG2560-0489
    48 mm
    Closure
    Connexion Pour la disponibilité
    1 * 4 ST - 262,00 €